Assay Design



Gene Transcripts

Gene Symbol
DDX39B
Entrez Gene ID
448813
Gene Name
DExD-box helicase 39B
Gene Aliases
BAT1
Chromosome Location
Chr.7: 23658089 - 23669095 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005157.1 NP_001005157.1 1 - 2 216 81
XM_005665792.3 XP_005665849.1 - 452 81
XM_005665793.3 XP_005665850.1 - 439 81
XM_005665794.3 XP_005665851.1 - 503 81
XM_021098341.1 XP_020954000.1 - 883 81
GenBank mRNA AK236289.1 - 2 - 3 390 81
AK238677.1 - 2 - 3 405 81
AK389641.1 - 2 - 3 500 81
AK389706.1 - 2 - 3 407 81
AK399598.1 - 2 - 3 407 81
EU282340.1 - 1 - 2 867 81

View Full Product Details