Assay Design



Gene Transcripts

Gene Symbol
SSTR2
Entrez Gene ID
497059
Gene Name
somatostatin receptor 2
Gene Aliases
-
Chromosome Location
Chr.12: 7783401 - 7789483 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011694.1 NP_001011694.1 1 - 1 1054 81
XM_005668643.3 XP_005668700.1 - 1493 81
GenBank mRNA AK390280.1 - 2 - 2 1395 81

View Full Product Details