Assay Design



Gene Transcripts

Gene Symbol
TNFSF10
Entrez Gene ID
406191
Gene Name
TNF superfamily member 10
Gene Aliases
TRAIL
Chromosome Location
Chr.13: 111021952 - 111103060 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024696.1 NP_001019867.1 3 - 4 334 129
XM_021068572.1 XP_020924231.1 - 620 129
GenBank mRNA AK394230.1 - 3 - 4 326 129
AY639873.1 - 3 - 4 334 129

View Full Product Details