Assay Design



Gene Transcripts

Gene Symbol
MTX1
Entrez Gene ID
664654
Gene Name
metaxin 1
Gene Aliases
Metaxin-1
Chromosome Location
Chr.4: 94606576 - 94611439 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039750.1 NP_001034839.1 1 - 2 95 115
GenBank mRNA AB239841.1 - 1 - 2 106 115
AK234275.1 - 1 - 2 106 115
AK347776.1 - 1 - 2 82 115
AK392795.1 - 1 - 2 99 115
DQ402035.1 - 1 - 2 95 115

View Full Product Details