Assay Design



Gene Transcripts

Gene Symbol
RLN2
Entrez Gene ID
396891
Gene Name
relaxin 2
Gene Aliases
RLN
Chromosome Location
Chr.1: 216763546 - 216769802 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213872.1 NP_999037.1 1 - 2 209 117
GenBank mRNA A06852.1 - - 209 117
A16593.1 - - 209 117
AK395920.1 - 1 - 2 316 117
K01088.1 - 1 - 2 209 117

View Full Product Details