Assay Design
Gene Transcripts
- Gene Symbol
- OVGP1
- Entrez Gene ID
-
397140
- Gene Name
- oviductal glycoprotein 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 108929034 - 108941040 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_214070.1 |
NP_999235.1 |
9 - 10 |
1027 |
95 |
GenBank mRNA |
U43490.1 |
- |
9 - 10 |
1027 |
95 |
View Full Product Details