Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_214110.1 | NP_999275.1 | 7 - 8 | 796 | 102 |
XM_005671145.3 | XP_005671202.1 | - | 1014 | 102 | |
XM_013983376.2 | XP_013838830.1 | - | 843 | 102 | |
GenBank mRNA | AF132496.2 | - | 7 - 8 | 796 | 102 |