Assay Design



Gene Transcripts

Gene Symbol
MGP
Entrez Gene ID
397206
Gene Name
matrix Gla protein
Gene Aliases
MPG
Chromosome Location
Chr.5: 57665272 - 57669360 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214116.1 NP_999281.1 3 - 4 217 60
GenBank mRNA AF525316.1 - 3 - 4 217 60
AK346558.1 - 3 - 4 236 60
GU391766.1 - 3 - 4 236 60

View Full Product Details