Assay Design



Gene Transcripts

Gene Symbol
TGFB3
Entrez Gene ID
397400
Gene Name
transforming growth factor beta 3
Gene Aliases
-
Chromosome Location
Chr.7: 99133873 - 99162239 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214198.1 NP_999363.1 3 - 4 910 64
XM_005666355.3 XP_005666412.1 - 1960 64
XM_013978322.2 XP_013833776.1 - 1690 64
GenBank mRNA AK236343.1 - 4 - 5 1941 64
AK240494.1 - 3 - 4 360 64
X14150.1 - - 910 64

View Full Product Details