Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_214198.1 | NP_999363.1 | 5 - 6 | 1186 | 55 |
XM_005666355.3 | XP_005666412.1 | - | 2236 | 55 | |
XM_013978322.2 | XP_013833776.1 | - | 1966 | 55 | |
GenBank mRNA | AK236343.1 | - | 6 - 7 | 2217 | 55 |
AK240494.1 | - | 5 - 6 | 636 | 55 | |
X14150.1 | - | - | 1186 | 55 |