Assay Design



Gene Transcripts

Gene Symbol
SFTPA1
Entrez Gene ID
397503
Gene Name
surfactant protein A1
Gene Aliases
PSAP, PSP-A, SFTPA, SP-A
Chromosome Location
Chr.14: 82091178 - 82095393 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348959.1 NP_001335888.1 3 - 4 380 106
GenBank mRNA EU622632.1 - 3 - 4 380 106
L41350.1 - 3 - 4 343 106

View Full Product Details