Assay Design



Gene Transcripts

Gene Symbol
IL13RA1
Entrez Gene ID
397615
Gene Name
interleukin 13 receptor subunit alpha 1
Gene Aliases
-
Chromosome Location
Chr.X: 97300960 - 97353754 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214341.1 NP_999506.1 10 - 11 1179 127
GenBank mRNA AK395683.1 - 10 - 11 1224 127
AY266142.1 - 10 - 11 1179 127

View Full Product Details