Assay Design
Gene Transcripts
- Gene Symbol
- HBE1
- Entrez Gene ID
-
407067
- Gene Name
- hemoglobin subunit epsilon 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 4818206 - 4820243 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_214447.1 |
NP_999612.1 |
1 - 2 |
149 |
75 |
View Full Product Details