Assay Design



Gene Transcripts

Gene Symbol
LTA
Entrez Gene ID
407742
Gene Name
lymphotoxin alpha
Gene Aliases
TNFB
Chromosome Location
Chr.7: 23696387 - 23698280 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214453.1 NP_999618.1 2 - 3 633 135
GenBank mRNA AK348835.1 - 3 - 4 576 135
EU282349.1 - 3 - 4 347 135

View Full Product Details