Assay Design



Gene Transcripts

Gene Symbol
SGCZ
Entrez Gene ID
100157832
Gene Name
sarcoglycan zeta
Gene Aliases
-
Chromosome Location
Chr.17: 2028093 - 3049613 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144844.1 NP_001138316.1 2 - 2 240 77
XM_021077238.1 XP_020932897.1 - 4476 77
XM_021077239.1 XP_020932898.1 - 2574 77
XM_021077240.1 XP_020932899.1 - 2139 77
XM_021077241.1 XP_020932900.1 - 1471 77
XM_021077242.1 XP_020932901.1 - 1346 77
GenBank mRNA FJ524379.1 - 2 - 2 240 77

View Full Product Details