Assay Design



Gene Transcripts

Gene Symbol
KPNA7
Entrez Gene ID
100302505
Gene Name
karyopherin subunit alpha 7
Gene Aliases
-
Chromosome Location
Chr.3: 6167382 - 6201282 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163411.1 NP_001156883.1 4 - 5 608 82
XM_021085626.1 XP_020941285.1 - 1042 82
GenBank mRNA AK348492.1 - 4 - 5 606 82
GQ166958.1 - 4 - 5 608 82

View Full Product Details