Assay Design



Gene Transcripts

Gene Symbol
HAVCR1
Entrez Gene ID
100303721
Gene Name
hepatitis A virus cellular receptor 1
Gene Aliases
-
Chromosome Location
Chr.16: 66162318 - 66208868 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164736.1 NP_001158208.1 6 - 7 922 86
XM_021076535.1 XP_020932194.1 - 1357 86
GenBank mRNA GQ421749.1 - 6 - 7 922 86

View Full Product Details