Assay Design



Gene Transcripts

Gene Symbol
RAG2
Entrez Gene ID
100151744
Gene Name
recombination activating 2
Gene Aliases
RAG-2
Chromosome Location
Chr.2: 24547605 - 24553944 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128481.1 NP_001121953.1 1 - 1 1453 95
XM_021076940.1 XP_020932599.1 - 1693 95

View Full Product Details