Assay Design



Gene Transcripts

Gene Symbol
CYP1A2
Entrez Gene ID
100152910
Gene Name
cytochrome P450, family 1, subfamily A, polypeptide 2
Gene Aliases
-
Chromosome Location
Chr.7: 58779529 - 58786155 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159614.1 NP_001153086.1 1 - 2 827 57
XM_005666124.3 XP_005666181.1 - 975 57
GenBank mRNA KM817029.1 - 2 - 3 920 57

View Full Product Details