Assay Design



Gene Transcripts

Gene Symbol
OTUB1
Entrez Gene ID
100302092
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 1
Gene Aliases
-
Chromosome Location
Chr.2: 8088528 - 8095555 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162403.1 NP_001155875.1 4 - 5 363 73
GenBank mRNA AK232154.1 - 4 - 5 360 73
AK392248.1 - 4 - 5 375 73
AK398824.1 - 4 - 5 350 73
AY609773.1 - 4 - 5 357 73
EU310930.1 - 4 - 5 363 73

View Full Product Details