Assay Design



Gene Transcripts

Gene Symbol
ODF3
Entrez Gene ID
100499231
Gene Name
outer dense fiber of sperm tails 3
Gene Aliases
-
Chromosome Location
Chr.2: 20869 - 25845 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348539.1 NP_001335468.1 3 - 4 446 71
XM_021077666.1 XP_020933325.1 - 1655 71
GenBank mRNA HM138511.1 - 3 - 4 71

View Full Product Details