Assay Design



Gene Transcripts

Gene Symbol
AGO4
Entrez Gene ID
100499507
Gene Name
argonaute RISC component 4
Gene Aliases
EIF2C4
Chromosome Location
Chr.6: 91967499 - 92013030 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194972.1 NP_001181901.1 17 - 18 2474 65
XM_021093284.1 XP_020948943.1 - 5451 65
XM_021093286.1 XP_020948945.1 - 2672 65
XM_021093287.1 XP_020948946.1 - 3897 65
GenBank mRNA GQ329717.1 - 17 - 18 2474 65

View Full Product Details