Assay Design



Gene Transcripts

Gene Symbol
AGO1
Entrez Gene ID
100499510
Gene Name
argonaute RISC component 1
Gene Aliases
EIF2C1
Chromosome Location
Chr.6: 92038104 - 92076477 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194976.1 NP_001181905.1 13 - 14 1740 77
XM_005665209.3 XP_005665266.1 - 1861 77
XM_021093293.1 XP_020948952.1 - 1451 77
GenBank mRNA AK344581.1 - 12 - 13 1640 77
GQ329714.1 - 13 - 14 1740 77

View Full Product Details