Assay Design



Gene Transcripts

Gene Symbol
AGO1
Entrez Gene ID
100499510
Gene Name
argonaute RISC component 1
Gene Aliases
EIF2C1
Chromosome Location
Chr.6: 92038104 - 92076477 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194976.1 NP_001181905.1 16 - 17 2161 70
XM_005665209.3 XP_005665266.1 - 2282 70
XM_021093293.1 XP_020948952.1 - 1872 70
GenBank mRNA GQ329714.1 - 16 - 17 2161 70

View Full Product Details