Assay Design



Gene Transcripts

Gene Symbol
VIP
Entrez Gene ID
100500718
Gene Name
vasoactive intestinal peptide
Gene Aliases
PHI
Chromosome Location
Chr.1: 13611786 - 13620250 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195233.1 NP_001182162.1 3 - 4 374 92
GenBank mRNA GU576974.1 - 3 - 4 374 92

View Full Product Details