Assay Design



Gene Transcripts

Gene Symbol
CMYA5
Entrez Gene ID
100505410
Gene Name
cardiomyopathy associated 5
Gene Aliases
-
Chromosome Location
Chr.2: 88391104 - 88506336 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197307.1 NP_001184236.1 10 - 11 11888 79
XM_021077698.1 XP_020933357.1 - 11943 79
GenBank mRNA AK394690.1 - 8 - 9 952 79
FJ208850.1 - 10 - 11 11888 79

View Full Product Details