Assay Design



Gene Transcripts

Gene Symbol
BZW1
Entrez Gene ID
100152909
Gene Name
basic leucine zipper and W2 domains 1
Gene Aliases
-
Chromosome Location
Chr.15: 104525900 - 104543795 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244493.1 NP_001231422.1 9 - 10 1049 74
XM_021074514.1 XP_020930173.1 - 1375 74
GenBank mRNA AK239661.1 - 9 - 10 1055 74
AK343867.1 - 9 - 10 1101 74
AK343897.1 - 7 - 8 896 74
AK345146.1 - 3 - 4 647 74
AK394609.1 - - 551 74
AY609907.1 - 9 - 10 1053 74

View Full Product Details