Assay Design



Gene Transcripts

Gene Symbol
SLC30A2
Entrez Gene ID
100170651
Gene Name
solute carrier family 30 member 2
Gene Aliases
-
Chromosome Location
Chr.6: 83478761 - 83484876 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139475.1 NP_001132947.1 3 - 4 647 76
GenBank mRNA AK238522.1 - 4 - 5 641 76
EU825192.2 - 3 - 4 647 76

View Full Product Details