Assay Design



Gene Transcripts

Gene Symbol
TNFRSF12A
Entrez Gene ID
100217390
Gene Name
TNF receptor superfamily member 12A
Gene Aliases
Fn14, TWEAKR
Chromosome Location
Chr.3: 39166288 - 39168294 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142839.2 NP_001136311.1 2 - 3 218 76
GenBank mRNA AY609823.1 - 2 - 3 212 76
EU417836.1 - 2 - 3 218 76

View Full Product Details