Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143718.1 | NP_001137190.1 | 3 - 4 | 333 | 102 |
XM_013982013.2 | XP_013837467.1 | - | 531 | 102 | |
XM_013982014.2 | XP_013837468.2 | - | 773 | 102 | |
XM_021068336.1 | XP_020923995.1 | - | 463 | 102 | |
GenBank mRNA | AK352498.1 | - | 3 - 4 | 348 | 102 |
AK399967.1 | - | 3 - 4 | 350 | 102 | |
FJ436393.1 | - | 3 - 4 | 333 | 102 |