Assay Design



Gene Transcripts

Gene Symbol
MGLL
Entrez Gene ID
100233193
Gene Name
monoglyceride lipase
Gene Aliases
-
Chromosome Location
Chr.13: 72480395 - 72568964 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143718.1 NP_001137190.1 5 - 6 585 62
XM_013982014.2 XP_013837468.2 - 1025 62
XM_021068336.1 XP_020923995.1 - 715 62
GenBank mRNA AK350271.1 - 3 - 4 327 62
AK352498.1 - 5 - 6 600 62
AK399967.1 - 5 - 6 602 62
FJ436393.1 - 5 - 6 585 62

View Full Product Details