Assay Design



Gene Transcripts

Gene Symbol
ROPN1
Entrez Gene ID
100359353
Gene Name
rhophilin associated tail protein 1
Gene Aliases
ROPN1B
Chromosome Location
Chr.13: 136395628 - 136428170 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172369.1 NP_001165840.1 5 - 6 777 86
XM_021068368.1 XP_020924027.1 - 801 86
XM_021068369.1 XP_020924028.1 - 866 86
GenBank mRNA AK240561.1 - 5 - 6 772 86
GU373671.1 - 5 - 6 777 86

View Full Product Details