Assay Design



Gene Transcripts

Gene Symbol
ODF3B
Entrez Gene ID
100517096
Gene Name
outer dense fiber of sperm tails 3B
Gene Aliases
-
Chromosome Location
Chr.5: 215972 - 217973 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206375.1 NP_001193304.1 3 - 4 362 112
XM_005653167.2 XP_005653224.1 - 465 112
XM_021091042.1 XP_020946701.1 - 728 112
GenBank mRNA EW465602.2 - 3 - 4 369 112

View Full Product Details