Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244899.1 | NP_001231828.1 | 2 - 3 | 540 | 70 |
XM_021072055.1 | XP_020927714.1 | - | 558 | 70 | |
XM_021072056.1 | XP_020927715.1 | - | 559 | 70 | |
GenBank mRNA | AK346287.1 | - | 2 - 3 | 540 | 70 |