Assay Design



Gene Transcripts

Gene Symbol
KCTD1
Entrez Gene ID
100514526
Gene Name
potassium channel tetramerization domain containing 1
Gene Aliases
-
Chromosome Location
Chr.6: 111048193 - 111237868 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244971.2 NP_001231900.2 2 - 3 2006 70
XM_005653406.3 XP_005653463.1 - 1113 70
XM_005653407.3 XP_005653464.1 - 209 70
XM_021093325.1 XP_020948984.1 - 1952 70
GenBank mRNA AK395469.1 - 2 - 3 605 70
AY610046.1 - 2 - 3 315 70

View Full Product Details