Assay Design



Gene Transcripts

Gene Symbol
INPP1
Entrez Gene ID
100514878
Gene Name
inositol polyphosphate-1-phosphatase
Gene Aliases
-
Chromosome Location
Chr.15: 94956535 - 94987286 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001245012.1 NP_001231941.1 5 - 6 877 71
XM_005672032.3 XP_005672089.1 - 1054 71
XM_005672033.3 XP_005672090.1 - 1289 71
XM_021074607.1 XP_020930266.1 - 3062 71
GenBank mRNA AK232018.1 - 5 - 6 877 71
AK351036.1 - 4 - 5 698 71

View Full Product Details