Assay Design



Gene Transcripts

Gene Symbol
IMMP1L
Entrez Gene ID
100514893
Gene Name
inner mitochondrial membrane peptidase subunit 1
Gene Aliases
-
Chromosome Location
Chr.2: 29230172 - 29305906 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001245014.1 NP_001231943.1 6 - 7 687 103
XM_005661048.3 XP_005661105.1 - 581 103
GenBank mRNA AK347172.1 - 6 - 7 687 103

View Full Product Details