Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001245976.1 | NP_001232905.1 | 4 - 5 | 488 | 90 |
XM_021068454.1 | XP_020924113.1 | - | 552 | 90 | |
XM_021068455.1 | XP_020924114.1 | - | 829 | 90 | |
XM_021068456.1 | XP_020924115.1 | - | 622 | 90 | |
XM_021068457.1 | XP_020924116.1 | - | 585 | 90 | |
XM_021068458.1 | XP_020924117.1 | - | 925 | 90 | |
XM_021068459.1 | XP_020924118.1 | - | 515 | 90 | |
GenBank mRNA | AK393600.1 | - | 4 - 5 | 483 | 90 |
JF271685.1 | - | 4 - 5 | 488 | 90 |