Assay Design



Gene Transcripts

Gene Symbol
THAP11
Entrez Gene ID
100520959
Gene Name
THAP domain containing 11
Gene Aliases
-
Chromosome Location
Chr.6: 28465458 - 28466387 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246182.1 NP_001233111.1 1 - 1 718 109
GenBank mRNA AK239211.1 - 1 - 1 983 109
AK396579.1 - 1 - 1 983 109
JF439088.1 - 1 - 1 718 109

View Full Product Details