Assay Design



Gene Transcripts

Gene Symbol
KCNE3
Entrez Gene ID
397168
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 3
Gene Aliases
-
Chromosome Location
Chr.9: 8810716 - 8822191 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214093.2 NP_999258.1 2 - 3 163 83
GenBank mRNA AK390470.1 - 2 - 3 162 83
FS646243.1 - 2 - 3 165 83

View Full Product Details