Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243375.1 | NP_001230304.1 | 1 - 2 | 90 | 71 |
XM_021093438.1 | XP_020949097.1 | - | 126 | 71 | |
GenBank mRNA | AK235444.1 | - | 1 - 2 | 92 | 71 |
AK348694.1 | - | 1 - 2 | 90 | 71 | |
FS693335.1 | - | 1 - 2 | 90 | 71 |