Assay Design



Gene Transcripts

Gene Symbol
IGSF6
Entrez Gene ID
100627993
Gene Name
immunoglobulin superfamily member 6
Gene Aliases
-
Chromosome Location
Chr.3: 23673162 - 23683256 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243387.1 NP_001230316.1 5 - 6 706 101
GenBank mRNA AK231345.1 - 5 - 6 706 101

View Full Product Details