Assay Design



Gene Transcripts

Gene Symbol
C5H12orf4
Entrez Gene ID
100153309
Gene Name
chromosome 5 C12orf4 homolog
Gene Aliases
-
Chromosome Location
Chr.5: 65896750 - 65944183 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243412.1 NP_001230341.1 9 - 10 1260 86
NM_001243413.1 NP_001230342.1 8 - 9 1117 86
XM_005655700.3 XP_005655757.1 - 1900 86
XM_013988430.2 XP_013843884.1 - 1985 86
XM_013988431.2 XP_013843885.1 - 1159 86
XM_021090919.1 XP_020946578.1 - 1906 86
XM_021090921.1 XP_020946580.1 - 1149 86
XM_021090922.1 XP_020946581.1 - 1985 86
XM_021090923.1 XP_020946582.1 - 974 86
XM_021090924.1 XP_020946583.1 - 1711 86
GenBank mRNA AK343836.1 - 9 - 10 1244 86
AK400126.1 - 9 - 10 1243 86
BP443627.1 - 5 - 6 645 86

View Full Product Details