Assay Design



Gene Transcripts

Gene Symbol
VPS4A
Entrez Gene ID
100627841
Gene Name
vacuolar protein sorting 4 homolog A
Gene Aliases
-
Chromosome Location
Chr.6: 17657792 - 17669560 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243418.1 NP_001230347.1 1 - 2 175 71
XM_005655767.3 XP_005655824.1 - 183 71
XM_021093432.1 XP_020949091.1 - 183 71
GenBank mRNA AK237952.1 - 1 - 2 176 71
AK239378.1 - 1 - 2 147 71
AK392132.1 - 1 - 2 171 71
AK398017.1 - 1 - 2 172 71

View Full Product Details