Assay Design



Gene Transcripts

Gene Symbol
PIGP
Entrez Gene ID
100154885
Gene Name
phosphatidylinositol glycan anchor biosynthesis class P
Gene Aliases
-
Chromosome Location
Chr.13: 200686716 - 200695537 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243520.1 NP_001230449.1 3 - 4 438 108
NM_001243521.1 NP_001230450.1 3 - 4 223 108
NM_001243522.1 NP_001230451.1 3 - 4 285 108
XM_021068231.1 XP_020923890.1 - 361 108
GenBank mRNA AK344057.1 - 3 - 4 232 108
AK392640.1 - 3 - 4 278 108
AK399784.1 - 3 - 4 434 108
BM484478.1 - 3 - 4 222 108
EW470448.2 - 3 - 4 285 108

View Full Product Details