Assay Design



Gene Transcripts

Gene Symbol
TDP2
Entrez Gene ID
100157860
Gene Name
tyrosyl-DNA phosphodiesterase 2
Gene Aliases
-
Chromosome Location
Chr.7: 19489381 - 19502437 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243686.1 NP_001230615.1 2 - 3 295 114
GenBank mRNA AK350141.1 - 2 - 3 295 114

View Full Product Details