Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243711.1 | NP_001230640.1 | 2 - 3 | 208 | 84 |
XM_005660986.2 | XP_005661043.1 | - | 505 | 84 | |
XM_005660987.3 | XP_005661044.1 | - | 348 | 84 | |
XM_021077723.1 | XP_020933382.1 | - | 197 | 84 | |
GenBank mRNA | AK349798.1 | - | 2 - 3 | 513 | 84 |
AK396456.1 | - | 2 - 3 | 202 | 84 | |
AK400493.1 | - | 2 - 3 | 296 | 84 | |
FS675912.1 | - | 2 - 3 | 220 | 84 | |
FS721678.1 | - | 2 - 3 | 208 | 84 |