Assay Design



Gene Transcripts

Gene Symbol
RBMX
Entrez Gene ID
100158042
Gene Name
RNA binding motif protein X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 111965859 - 111974544 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244096.1 NP_001231025.1 2 - 3 243 81
XM_013986455.2 XP_013841909.1 - 298 81
XM_021079411.1 XP_020935070.1 - 315 81
XM_021079412.1 XP_020935071.1 - 298 81
GenBank mRNA AK235135.1 - 2 - 3 243 81
AK236383.1 - 2 - 3 262 81
AK236451.1 - 2 - 3 246 81
AK236585.1 - 2 - 3 248 81
AK394814.1 - 2 - 3 81
AK397739.1 - 2 - 3 298 81

View Full Product Details