Assay Design



Gene Transcripts

Gene Symbol
SMAD7
Entrez Gene ID
100521305
Gene Name
SMAD family member 7
Gene Aliases
-
Chromosome Location
Chr.1: 98512800 - 98543924 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244175.1 NP_001231104.1 2 - 3 669 71
GenBank mRNA EW502633.2 - 1 - 2 53 71

View Full Product Details