Assay Design



Gene Transcripts

Gene Symbol
CHCHD3
Entrez Gene ID
414383
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Gene Aliases
-
Chromosome Location
Chr.18: 16052611 - 16348998 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244176.1 NP_001231105.1 5 - 6 620 108
XM_021078525.1 XP_020934184.1 - 642 108
XM_021078526.1 XP_020934185.1 - 648 108
XM_021078527.1 XP_020934186.1 - 642 108
XM_021078528.1 XP_020934187.1 - 642 108
XM_021078529.1 XP_020934188.1 - 641 108
XM_021078530.1 XP_020934189.1 - 648 108
GenBank mRNA AK231938.1 - 5 - 6 605 108
AK234688.1 - 5 - 6 557 108
AK348653.1 - 5 - 6 559 108
AK393133.1 - 5 - 6 608 108
AK398750.1 - 5 - 6 634 108
AY610152.1 - 5 - 6 620 108

View Full Product Details